78482

Sigma-Aldrich

HypoGel® 200 OH

extent of labeling: ~0.8 mmol/g loading

Synonym(s):
O-(2-Hydroxyethyl)tetra(oxyethylene) polymer-bound
Pricing and availability is not currently available.

extent of labeling

~0.8 mmol/g loading

particle size

110-150 μm

storage temp.

2-8°C

Legal Information

HypoGel is a registered trademark of Rapp Polymere GmbH

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

WGK Germany

3

Certificate of Analysis
Certificate of Origin
Sören Twarock et al.
Molecular cancer, 10, 30-30 (2011-03-25)
Oesophageal cancer is a highly aggressive tumour entity with at present poor prognosis. Therefore, novel treatment options are urgently needed. Hyaluronan (HA) is a polysaccharide present in the matrix of human oesophageal squamous cell carcinoma (ESCC). Importantly, in vitro ESCC...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.