80067

Supelco

Amyl hexanoate

analytical standard

Synonym(s):
n-Pentyl hexanoate, Amyl caproate, NSC 53794, Pentyl caproate, Hexanoic acid pentyl ester, NSC 46119
Linear Formula:
CH3(CH2)4CO2(CH2)4CH3
CAS Number:
Molecular Weight:
186.29
EC Number:
MDL number:
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Quality Level

grade

analytical standard

assay

≥98.5% (GC)

form

neat

shelf life

limited shelf life, expiry date on the label

application(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.418-1.422
n20/D 1.42 (lit.)

bp

226 °C (lit.)

density

0.858 g/mL at 25 °C (lit.)

Featured Industry

Cleaning Products
Cosmetics
Flavors and Fragrances
Food and Beverages
Personal Care

format

neat

SMILES string

CCCCCOC(=O)CCCCC

InChI

1S/C11H22O2/c1-3-5-7-9-11(12)13-10-8-6-4-2/h3-10H2,1-2H3

InChI key

WRFZKAGPPQGDDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

RIDADR

NA 1993 / PGIII

WGK Germany

WGK 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.