All Photos(1)

89880

Sigma-Aldrich

o-Toluic acid

puriss., ≥99.5% (T)

Synonym(s):
2-Methylbenzoic acid
Linear Formula:
CH3C6H4CO2H
CAS Number:
Molecular Weight:
136.15
Beilstein:
1072103
EC Number:
MDL number:
PubChem Substance ID:

grade

puriss.

assay

≥99.5% (T)

ign. residue

≤0.05%

bp

258-259 °C (lit.)

mp

102-104 °C (lit.)
103-105 °C

density

1.062 g/mL at 25 °C (lit.)

SMILES string

Cc1ccccc1C(O)=O

InChI

1S/C8H8O2/c1-6-4-2-3-5-7(6)8(9)10/h2-5H,1H3,(H,9,10)

InChI key

ZWLPBLYKEWSWPD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

K H Wang et al.
Chemosphere, 40(4), 389-394 (2000-02-09)
This investigation used UV light of 365 nm and titanium dioxide in aqueous suspension to study the photocatalytic reaction of o-methylbenzoic acid under the influence of pH values, anion additives and the varieties of titanium dioxide. From experimental results, under...
D Durkin et al.
Electrophoresis, 21(10), 1997-2009 (2000-07-06)
The concept of dual opposite injection in capillary electrophoresis (DOI-CE) for the simultaneous separation, under conditions of suppressed electroosmotic flow, of anionic and cationic compounds with no bias in resolution and analysis time, is extended to a higher pH range...
Shiu-Mei Liu et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 38(6), 1099-1113 (2003-05-31)
Composition of the headspace gas affected the biotransformation pattern of toluic acid isomers in anoxic sediment slurries. Under an N2 atmosphere, o- and m-, and p-toluic acid (20-25 mg L(-1)) were biotransformed in 100 days, 77 days, and 148 days...
K Watanabe et al.
Drug metabolism and disposition: the biological fate of chemicals, 23(2), 261-265 (1995-02-01)
Mouse hepatic microsomal enzymes catalyzed the oxidation of o-, m-, and p-tolualdehydes, intermediate metabolites of xylene, to the corresponding toluic acids. Cofactor requirement for the catalytic activity indicates that the microsomes contain NAD- and NADPH-dependent enzymes for this reaction. GC/MS...
Paula Esquivias et al.
Oncology reports, 31(6), 2785-2791 (2014-04-17)
Aspirin has been proposed in recent years as a candidate for chemoprevention of adenocarcinoma in patients with Barrett's esophagus. The aim of the present study was to evaluate the effect of acetylsalicylic acid (ASA) in an experimental model of esophageal...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service