90102

Supelco

17β(H),21α(H)-30-Norhopane solution

0.1 mg/mL in isooctane, analytical standard

Synonym(s):
21α(H)-30-nor-A′-neogammacerane
Empirical Formula (Hill Notation):
C29H50
CAS Number:
Molecular Weight:
398.71
EC Number:
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

grade

analytical standard

assay

≥97.0% (GC/MS)

shelf life

limited shelf life, expiry date on the label

concentration

0.1 mg/mL in isooctane

application(s)

HPLC: suitable
gas chromatography (GC): suitable

Featured Industry

Food and Beverages

format

single component solution

storage temp.

2-8°C

SMILES string

HC@@12CCC@3(C)C@(H)(CCC@4(H)C@@5(C)CCCC(C)(C)C@5(H)CCC@@34C)C@@1(C)CCC@H2CC

InChI

1S/C29H50/c1-8-20-12-17-26(4)21(20)13-18-28(6)23(26)10-11-24-27(5)16-9-15-25(2,3)22(27)14-19-29(24,28)7/h20-24H,8-19H2,1-7H3/t20-,21+,22+,23-,24-,26+,27+,28-,29-/m1/s1

InChI key

XKJROQIFLGXWEY-LMDOITEXSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

F,Xn,N

Risk Statement

11-38-50/53-65-67

Safety Statement

9-16-29-33-60-61-62

RIDADR

UN1262 - class 3 - PG 2 - Octanes, solution

WGK Germany

2

Flash Point(F)

10.4 °F

Flash Point(C)

-12 °C

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.