16590

Sigma-Aldrich

Atto 520 maleimide

for fluorescence, ≥90% (coupling rate)

Empirical Formula (Hill Notation):
C28H33ClN4O8
Molecular Weight:
589.04
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

grade

for fluorescence

assay

≥90% (coupling rate)

solubility

DMF: soluble
DMSO: soluble
acetonitrile: soluble

suitability

passes test for fluorescence

storage temp.

−20°C

SMILES string

O-Cl(=O)(=O)=O.CCNc1cc2OC3=CC(=NH+\CC)\C(C)=CC3=C(CCC(=O)NCCN4C(=O)C=CC4=O)c2cc1C

InChI

1S/C28H32N4O4.ClHO4/c1-5-29-22-15-24-20(13-17(22)3)19(21-14-18(4)23(30-6-2)16-25(21)36-24)7-8-26(33)31-11-12-32-27(34)9-10-28(32)35;2-1(3,4)5/h9-10,13-16,29H,5-8,11-12H2,1-4H3,(H,31,33);(H,2,3,4,5)/b30-23-;

InChI key

DGEQWOHLXNEPEL-KZJPCHATSA-N

Application

Atto 520 maleimide utilized for labeling sulfhydryl (thiol) groups of proteins, especially cysteine residues, to fluorescently study changes in conformation, binding, and kinetics .
Atto fluorescent labels are designed for high sensitivity applications, including single molecule detection. Atto labels have rigid structures that do not show any cis-trans-isomerization. Thus these labels display exceptional intensity with minimal spectral shift on conjugation.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Manoj Kumbhakar et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 10(4), 629-633 (2009-01-30)
Complex dynamics of combined long-range fluorescence resonance energy transfer (FRET) and short-range electron transfer (ET) processes in a single double-stranded DNA (dsDNA) molecule (see figure) reveals that FRET remains almost unaltered in the presence of ET. Present systems also demonstrate...
Vladimir Mekler et al.
The Journal of biological chemistry, 286(1), 270-279 (2010-10-19)
Promoter recognition by RNA polymerase is a key point in gene expression and a target of regulation. Bacterial RNA polymerase binds promoters in the form of the holoenzyme, with the σ specificity subunit being primarily responsible for promoter recognition. Free...
Hee-Jin Jeong et al.
Biosensors & bioelectronics, 40(1), 17-23 (2012-07-17)
Protein phosphorylation is a key event in intracellular signal transduction, and fluorescent biosensor for the specific phosphorylation event in a target protein is considered highly useful as a tool of cellular biology and drug screening. Vimentin, the most abundant intermediate...
Christopher A Teske et al.
The journal of physical chemistry. B, 109(28), 13811-13817 (2006-07-21)
Confocal laser scanning microscopy (CLSM) is being increasingly used for observing protein uptake in porous chromatography resins. Recent CLSM studies have revealed the possible existence of a nondiffusive protein transport mechanism. Observing protein uptake with CLSM requires labeling the protein...
Siddharth Nanguneri et al.
PloS one, 7(5), e38098-e38098 (2012-06-05)
Three-dimensional fluorescence imaging of thick tissue samples with near-molecular resolution remains a fundamental challenge in the life sciences. To tackle this, we developed tomoSTORM, an approach combining single-molecule localization-based super-resolution microscopy with array tomography of structurally intact brain tissue. Consecutive...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.