MilliporeSigma
All Photos(1)

316032

Sigma-Aldrich

Dibutyl phthalate-(carbonyl-14C)

≥98% (HPLC), toluene solution

Synonym(s):
Phthalic acid dibutyl ester-(carbonyl-14C)
Linear Formula:
C6H4-1,2-[14CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
278.34
MDL number:
PubChem Substance ID:

assay

≥98% (HPLC)

form

toluene solution

extent of labeling

10-30 mCi per mmol

application(s)

agriculture
environmental

mass shift

M+4

storage temp.

2-8°C

SMILES string

CCCCO[14C](=O)c1ccccc1[14C](=O)OCCCC

Looking for similar products? Visit Product Comparison Guide

Packaging

sealed ampule

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service