MilliporeSigma
All Photos(1)

322679

Sigma-Aldrich

Methyl iodide-14C

≥95% radiochemical purity basis (GC, HPLC), liquid

Synonym(s):
Methyl-14C iodide
Linear Formula:
14CH3I
CAS Number:
Molecular Weight:
141.94
MDL number:

assay

≥95% radiochemical purity basis (GC, HPLC)

form

liquid

extent of labeling

40-60 mCi per mmol

mass shift

M+2

storage temp.

2-8°C

SMILES string

[14CH3]I

Looking for similar products? Visit Product Comparison Guide

Packaging

Break-seal vial

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service