63401-011

Sigma-Aldrich

STRboost®

300 rxn, for multiplexing PCR samples

Pricing and availability is not currently available.

form

dry

usage

300 reactions

storage temp.

room temp

General description

STRboost® is a novel reagent specifically formulated to enhance multiplex PCR amplification reactions used for human identification applications. STRboost® can overcome limitations associated with multiplex STR analysis performed on compromised sample types. Use of STRboost® resolves stochastic effects such as peak balance issues, allele dropout and sub-optimal peak height definition.

Don′t see what you are looking for? Custom stabilization services for molecular diagnostic assay and reagent manufacturers are available through Sigma′s partner,
Biomatrica. PCRstable® enables the preservation of PCR-based assay components in any format (liquid or dry).

Packaging

1 bottle

Physical form

300 reactions (1 mL) in 1.5 mL screw-top microfuge tube made of durable polypropylene.

Legal Information

PCRstable is a registered trademark of Biomatrica
STRboost is a registered trademark of Biomatrica

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.