79405

Sigma-Aldrich

3-sn-Phosphatidyl-L-serine solution

from bovine brain, ≥98.0% (TLC)

Synonym(s):
L-α-Phosphatidyl-L-serine, PS
Beilstein/REAXYS Number:
1730695

biological source

bovine brain

assay

≥98.0% (TLC)

composition

18:0, ~40% (fatty acid)
18:1, ~28% (fatty acid)

concentration

10 mg/mL in chloroform: methanol (19:1)

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Other Notes

Sales restrictions may apply

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service