89740

Supelco

Atto 390 maleimide

BioReagent, suitable for fluorescence, ≥80% (coupling rate)

MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

Quality Level

product line

BioReagent

assay

≥80% (coupling rate)

fluorescence

λex 390 nm; λem 472 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

CC1CC(C)(C)N(CCCC(=O)NCCN2C(=O)C=CC2=O)c3cc4OC(=O)C=C(C)c4cc13

InChI

1S/C26H31N3O5/c1-16-12-25(33)34-21-14-20-18(13-19(16)21)17(2)15-26(3,4)29(20)10-5-6-22(30)27-9-11-28-23(31)7-8-24(28)32/h7-8,12-14,17H,5-6,9-11,15H2,1-4H3,(H,27,30)

InChI key

HHEQCRHPOHPYGI-UHFFFAOYSA-N

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

3

Qiang Li et al.
The journal of physical chemistry. B, 115(46), 13643-13649 (2011-10-15)
Fluorescence resonance energy transfer (FRET) using biotinylated β-galactosidase (βGAL) as a donor and Alexa Fluor 350 (AF350) labeled avidin as an acceptor has been investigated by means of steady-state fluorescence and time-resolved fluorescence spectroscopy. The donors are readily paired with...
Cuifang Kuang et al.
The Review of scientific instruments, 81(5), 053709-053709 (2010-06-03)
Stimulated emission depletion (STED) microscopy is one of the breakthrough technologies that belong to far-field optical microscopy and can achieve nanoscale spatial resolution. We demonstrate a far-field optical nanoscopy based on continuous wave lasers with different wavelengths, i.e., violet and...
Simon Corrie et al.
Molecular bioSystems, 5(3), 262-268 (2009-02-20)
Herein we report a method for the detection of methylated CpG dinucleotides located within CpG islands in genomic DNA using multiplexed bead-based assays and standard flow cytometry instrumentation. Four CpG "clusters" were identified in the TFPI2 and SPARC CpG islands...
Philip Howes et al.
Journal of the American Chemical Society, 132(11), 3989-3996 (2010-02-24)
Semiconducting polymer nanospheres (SPNs) have been synthesized and encapsulated in phospholipid micelles by a solvent evaporation technique. Four different conjugated polymers were used, yielding aqueous dispersions of nanoparticles which emit across the visible spectrum. The synthesis was simple and easily...
Gerhild Zauner et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 13(25), 7085-7090 (2007-06-20)
A fluorescence-based system to sense oxygen in solution is described. The method exploits the sensitivity of the endogenous fluorescence of type-3 copper proteins towards the presence of oxygen by translating the near-UV emission of the protein to label fluorescence in...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.