93121-017

Sigma-Aldrich

CloneStable®

greener alternative

Tube Kit, for room temp preservation of DNA

Pricing and availability is not currently available.

grade

for molecular biology

form

dry

usage

75 reactions

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

storage temp.

room temp

General description

SampleMatrix® by Biomatrica® is a proprietary core technology to stabilize and protect biological materials at room temperature without degradation. The SampleMatrix technology was designed by combining extremophile biology that enables long-term survival of organisms in extremely dry environments, with synthetic chemistry.

CloneStable® archives unpurified bacterial plasmid and genomic DNA at room temperature for long-term storage and shipping. Provided as a coating at the bottom of each tube or well, CloneStable® is completely dissolvable to ensure total sample recovery.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product was Designed for Energy Efficiency. To see how Stanford incorporated Biomatrica Technology and saved substantial amounts of energy, see their published study:

Biomatrica technology enables safe storage of biological material at room temperature.

Features and Benefits

  • Protects the integrity of bacterial DNA
  • Zero sample degradation
  • Store samples at room temperature

Packaging

  • Ideal format for shipping sample
  • Standard, snaplock 1.7 mL microtube made of durable polypropylene
  • Barcode labeled

Legal Information

Biomatrica is a registered trademark of Biomatrica
CloneStable is a registered trademark of Biomatrica
SampleMatrix is a registered trademark of Biomatrica

RIDADR

NONH for all modes of transport

WGK Germany

3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.