MilliporeSigma
All Photos(1)

A0259

Sigma-Aldrich

Acetarsone

≥99%

Synonym(s):
N-Acetyl-4-hydroxy-m-arsanilic acid
Empirical Formula (Hill Notation):
C8H10AsNO5
CAS Number:
Molecular Weight:
275.09
EC Number:
MDL number:
PubChem Substance ID:

assay

≥99%

SMILES string

CC(=O)Nc1cc(ccc1O)[As](O)(O)=O

InChI

1S/C8H10AsNO5/c1-5(11)10-7-4-6(9(13,14)15)2-3-8(7)12/h2-4,12H,1H3,(H,10,11)(H2,13,14,15)

InChI key

ODFJOVXVLFUVNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service