MilliporeSigma
All Photos(1)

A2004

Sigma-Aldrich

(±)-3-Aminobutyric acid

≥95%

Synonym(s):
β-Aminobutyric acid
Linear Formula:
CH3CH(NH2)CH2COOH
CAS Number:
Molecular Weight:
103.12
Beilstein:
1720563
EC Number:
MDL number:

assay

≥95%

SMILES string

CC(N)CC(O)=O

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service