A7259

Sigma-Aldrich

Aminoguanidine bicarbonate salt

Synonym(s):
Guanylhydrazine bicarbonate salt
Linear Formula:
NH2NHC(=NH)NH2 · H2CO3
CAS Number:
Molecular Weight:
136.11
Beilstein/REAXYS Number:
3569869
EC Number:
MDL number:
Pricing and availability is not currently available.

SMILES string

OC(O)=O.NNC(N)=N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibits both constitutive and inducible nitric oxide synthetase

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

Xi,N

Risk Statement

43-51/53

Safety Statement

36/37

RIDADR

UN 3077 9 / PGIII

WGK Germany

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.