A8835

Sigma-Aldrich

Aminoguanidine hydrochloride

≥98%

Synonym(s):
Guanylhydrazine hydrochloride
Linear Formula:
NH2NHC(=NH)NH2 · HCl
CAS Number:
Molecular Weight:
110.55
Beilstein/REAXYS Number:
3909606
MDL number:
Pricing and availability is not currently available.

assay

≥98%

mp

162-166 °C (lit.)

SMILES string

Cl.NNC(N)=N

InChI

1S/CH6N4.ClH/c2-1(3)5-4;/h4H2,(H4,2,3,5);1H

InChI key

UBDZFAGVPPMTIT-UHFFFAOYSA-N

Gene Information

human ... NOS1(4842), NOS2(4843), NOS2B(201288), NOS2C(645740), NOS3(4846)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibits both constitutive and inducible nitric oxide synthetase.

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®1280), a biologically annotated collection of high-quality, ready-to-screen compounds. Click here to learn more.

Legal Information

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.