A9808

Sigma-Aldrich

Pituitary adenylate cyclase activating polypeptide-27 ovine

≥97% (HPLC), solid

Synonym(s):
PACAP-27
Empirical Formula (Hill Notation):
C142H224N40O39S
CAS Number:
Molecular Weight:
3147.61
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

Quality Level

assay

≥97% (HPLC)

form

solid

composition

Peptide content, ≥65%

color

white

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)CNC(=O)C(CC(O)=O)NC(=O)C(CO)NC(=O)C(N)Cc1cnc[nH]1)C(=O)NC(Cc2ccccc2)C(=O)NC(C(C)O)C(=O)NC(CC(O)=O)C(=O)NC(CO)C(=O)NC(Cc3ccc(O)cc3)C(=O)NC(CO)C(=O)NC(CCCNC(N)=N)C(=O)NC(Cc4ccc(O)cc4)C(=O)NC(CCCNC(N)=N)C(=O)NC(CCCCN)C(=O)NC(CCC(N)=O)C(=O)NC(CCSC)C(=O)NC(C)C(=O)NC(C(C)C)C(=O)NC(CCCCN)C(=O)NC(CCCCN)C(=O)NC(Cc5ccc(O)cc5)C(=O)NC(CC(C)C)C(=O)NC(C)C(=O)NC(C)C(=O)NC(C(C)C)C(=O)NC(CC(C)C)C(N)=O

InChI

1S/C142H224N40O39S/c1-16-75(10)113(179-108(191)66-156-120(201)102(63-109(192)193)173-135(216)104(67-183)176-119(200)88(146)62-84-65-153-70-157-84)139(220)174-101(58-80-28-18-17-19-29-80)133(214)182-114(79(14)186)140(221)175-103(64-110(194)195)132(213)178-106(69-185)136(217)172-100(61-83-39-45-87(189)46-40-83)131(212)177-105(68-184)134(215)164-93(34-27-54-155-142(151)152)126(207)170-98(59-81-35-41-85(187)42-36-81)129(210)163-92(33-26-53-154-141(149)150)124(205)161-89(30-20-23-50-143)122(203)165-94(47-48-107(147)190)127(208)166-95(49-55-222-15)121(202)159-78(13)118(199)180-111(73(6)7)137(218)167-91(32-22-25-52-145)123(204)162-90(31-21-24-51-144)125(206)171-99(60-82-37-43-86(188)44-38-82)130(211)169-97(57-72(4)5)128(209)160-76(11)116(197)158-77(12)117(198)181-112(74(8)9)138(219)168-96(115(148)196)56-71(2)3/h17-19,28-29,35-46,65,70-79,88-106,111-114,183-189H,16,20-27,30-34,47-64,66-69,143-146H2,1-15H3,(H2,147,190)(H2,148,196)(H,153,157)(H,156,201)(H,158,197)(H,159,202)(H,160,209)(H,161,205)(H,162,204)(H,163,210)(H,164,215)(H,165,203)(H,166,208)(H,167,218)(H,168,219)(H,169,211)(H,170,207)(H,171,206)(H,172,217)(H,173,216)(H,174,220)(H,175,221)(H,176,200)(H,177,212)(H,178,213)(H,179,191)(H,180,199)(H,181,198)(H,182,214)(H,192,193)(H,194,195)(H4,149,150,154)(H4,151,152,155)

InChI key

RZGBUJXSKLDAFE-UHFFFAOYSA-N

Gene Information

human ... ADCY1(107)
mouse ... ADCY1(432530)
rat ... ADCY1(305509)
sheep ... PACAP(443337)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

His-Ser-Asp-Gly-Ile-Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-NH2

Biochem/physiol Actions

Neuropeptide that stimulates both adenylate cyclase in cultured pituitary cells and insulin release from pancreatic islet β-cells. Increases cAMP levels; increases tyrosine hydroxylase expression in chromaffin cells.

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
Emi Nakajima et al.
Molecular vision, 19, 174-183 (2013-02-05)
Our previous studies in the rabbit trigeminal nerve (TgN) showed that pituitary adenylate cyclase-activating peptide (PACAP) accelerated the extension of neuronal processes and recovery of corneal sensitivity. The purposes of the present study were 1) develop a procedure to culture...
Angela J Okragly et al.
Cephalalgia : an international journal of headache, 38(9), 1564-1574 (2017-11-07)
Background Many patients with migraines suffer from allergies and vice versa, suggesting a relationship between biological mechanisms of allergy and migraine. It was proposed many years ago that mast cells may be involved in the pathophysiology of migraines. We set...
Christiane Rose et al.
BMC neuroscience, 13, 84-84 (2012-07-25)
Amyloid precursor protein (APP), a key molecule in Alzheimer's disease (AD), is metabolized in two alternative cleavages, generating either the amyloidogenic peptides involved in AD pathology or the soluble form of APP (sAPPα). The level of amyloidogenic peptides in human...
B Koch et al.
Regulatory peptides, 38(1), 45-53 (1992-03-05)
The present study was aimed at investigating whether PACAP stimulates accumulation of cAMP, as well as hormonal secretion of homogeneous populations of pituitary proopiomelanocortin (POMC) cells, namely melanotrophs and AtT-20 corticotrophs. PACAP was shown to enhance cAMP accumulation in a...
S R Rawlings et al.
The Journal of biological chemistry, 269(8), 5680-5686 (1994-02-25)
Pituitary adenylate cyclase-activating polypeptide (PACAP) increases cAMP production and stimulates hormone release from a variety of anterior pituitary cells. However, in anterior pituitary gonadotrophs PACAP stimulates oscillations in cytosolic free Ca2+ concentration ([Ca2+]i) that appear to be independent of cAMP....

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.