ACE100

Sigma-Aldrich

Cultured Cells Acellularization Kit

Pricing and availability is not currently available.

Application

The Cultured Cells Acellularization Kit is for ECM preparation to be used as a supportive layer for cell growth, and for other applications like ECM characterization by immunofluorescence labeling or by Western blot analysis. This kit provides all of the reagents required for an easy and efficient removal of ECM-producing cultured cells (acellularization) leaving the underlying ECM intact. Additionally, it contains Ponceau S Solution for ECM staining and Solubilization Buffer for ECM protein extraction for downstream applications.
The kit was qualified with bovine aorta endothelial cells (BAEC), human foreskin fibroblasts, rabbit corneal primary cells, and with the NIH 3T3 cell line. Sufficient reagents are provide to process thirty 10 cm culture dishes.

Kit Components Also Available Separately

Product No.
Description
SDS

  • P7170Ponceau S solution, BioReagent, suitable for electrophoresis, 0.1 % (w/v) in 5% acetic acid 100 mL

  • P7170Ponceau S solution, BioReagent, suitable for electrophoresis, 0.1 % (w/v) in 5% acetic acid 30 mL

Hazard Codes

Xi

Risk Statement

36/37/38

Safety Statement

26-36

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.