All Photos(1)

B0379

Sigma-Aldrich

N-Benzoyl-DL-alanine

Empirical Formula (Hill Notation):
C10H11NO3
CAS Number:
Molecular Weight:
193.20
EC Number:
MDL number:

storage temp.

−20°C

SMILES string

CC(NC(=O)c1ccccc1)C(O)=O

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service