All Photos(1)

B3513

Sigma-Aldrich

Boc-N-Methyl-D-phenylalanine (dicyclohexylammonium) salt

Empirical Formula (Hill Notation):
C15H21NO4 · C12H23N
CAS Number:
Molecular Weight:
460.65
MDL number:

storage temp.

−20°C

SMILES string

C1CCC(CC1)NC2CCCCC2.CN([C@H](Cc3ccccc3)C(O)=O)C(=O)OC(C)(C)C

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service