B3782

Sigma-Aldrich

Restriction Enzyme Buffer SL

10 ×

Synonym(s):
Restriction Endonuclease Buffer
Pricing and availability is not currently available.

description

10× buffer contains 100 mM Tris-HCl, pH 7.5 at 37°C, 100 mM MgCl2, 10 mM DTE

concentration

10 ×

storage temp.

−20°C

SMILES string

CC(C)CC(CNC(Cc1ccccc1)C(N)=O)NC(=O)C(Cc2cncnH2)NC(=O)CNC(=O)C(NC(=O)C(C)NC(=O)C(Cc3cnHc4ccccc34)NC(=O)C(CCC(N)=O)NC(=O)C(N)Cc5cnHc6ccccc56)C(C)C

InChI

1S/C58H77N15O9/c1-32(2)21-39(29-65-46(52(61)76)22-35-13-7-6-8-14-35)69-57(81)48(25-38-28-62-31-67-38)70-50(75)30-66-58(82)51(33(3)4)73-53(77)34(5)68-56(80)47(24-37-27-64-44-18-12-10-16-41(37)44)72-55(79)45(19-20-49(60)74)71-54(78)42(59)23-36-26-63-43-17-11-9-15-40(36)43/h6-18,26-28,31-34,39,42,45-48,51,63-65H,19-25,29-30,59H2,1-5H3,(H2,60,74)(H2,61,76)(H,62,67)(H,66,82)(H,68,80)(H,69,81)(H,70,75)(H,71,78)(H,72,79)(H,73,77)

InChI key

YNZJHLFHFNCZIE-UHFFFAOYSA-N

General description

This buffer is for use with Sigma Restriction Enzymes. Please note every restriction enzyme is supplied with the appropriate buffer - this product is available for users who need additional buffer. For details on buffer composition and protocols for use, see the "Protocols and Articles" tab above.

RIDADR

NONH for all modes of transport

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.