All Photos(1)

B6888

Sigma-Aldrich

N6-Benzoyl-5′-O-(4,4′-dimethoxytrityl)-2′-deoxyadenosine 3′-(2-chlorophenyl) phosphate triethylammonium salt

~98%

Empirical Formula (Hill Notation):
C44H39ClN5O9P · C6H15N
CAS Number:
Molecular Weight:
949.43
EC Number:
MDL number:

assay

~98%

storage temp.

−20°C

SMILES string

CCN(CC)CC.COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(O)(=O)Oc3ccccc3Cl)n4cnc5c(NC(=O)c6ccccc6)ncnc45)(c7ccccc7)c8ccc(OC)cc8

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service