Documents

B8292

Sigma-Aldrich

5-(Benzyloxy)indole

powder

Synonym(s):
NSC 62895
Empirical Formula (Hill Notation):
C15H13NO
CAS Number:
Molecular Weight:
223.27
Beilstein/REAXYS Number:
173532
EC Number:
MDL number:
Pricing and availability is not currently available.

form

powder

impurities

≤7% solvents

color

white to light yellow

mp

100-104 °C (lit.)

solubility

ethanol: may be hazy yellow

storage temp.

2-8°C

SMILES string

C(Oc1ccc2nHccc2c1)c3ccccc3

InChI

1S/C15H13NO/c1-2-4-12(5-3-1)11-17-14-6-7-15-13(10-14)8-9-16-15/h1-10,16H,11H2

InChI key

JCQLPDZCNSVBMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Reactant in regio- and stereoselective morpholine-catalyzed direct C-3 alkenylation with α,β-unsaturated aldehydes
  • Reactant in selective debenzylation of protective groups using SiliaCat-palladium under mild reaction conditions
  • Reactant in metal-free Friedel-Crafts alkylation reactions
  • Reactant in preparation of protein kinase (PKC) inhibitors
  • Reactant in preparation of indole/quinoline carbothioic acid amide derivatives

Hazard Codes

Xi

Risk Statement

36/37/38

Safety Statement

26

WGK Germany

2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.