All Photos(1)

B9017

Sigma-Aldrich

Boc-Cys(Mbzl)-OH

Synonym(s):
Boc-S-(4-methylbenzyl)-L-cysteine
Linear Formula:
CH3C6H4CH2SCH2CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
325.42
Beilstein:
3594717
MDL number:

storage temp.

−20°C

SMILES string

Cc1ccc(CSC[C@H](NC(=O)OC(C)(C)C)C(O)=O)cc1

InChI

1S/C16H23NO4S/c1-11-5-7-12(8-6-11)9-22-10-13(14(18)19)17-15(20)21-16(2,3)4/h5-8,13H,9-10H2,1-4H3,(H,17,20)(H,18,19)/t13-/m0/s1

InChI key

CUNVVZWSABRKAL-ZDUSSCGKSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service