All Photos(1)

C1053

Supelco

Ethyl chrysanthemate

~95% (GC), Mixture of cis and trans isomers

Synonym(s):
2,2-Dimethyl-3-(1-isobutenyl)cyclopropane-1-carboxylic acid ethyl ester, Chrysanthemum monocarboxylic acid ethyl ester
Empirical Formula (Hill Notation):
C12H20O2
CAS Number:
Molecular Weight:
196.29
Beilstein:
2045740
EC Number:
MDL number:

assay

~95% (GC)

refractive index

n20/D 1.461 (lit.)

bp

112 °C/10 mmHg (lit.)

density

0.906 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)C1C(\C=C(\C)C)C1(C)C

InChI

1S/C12H20O2/c1-6-14-11(13)10-9(7-8(2)3)12(10,4)5/h7,9-10H,6H2,1-5H3

InChI key

VIMXTGUGWLAOFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A mixture of cis and trans isomers.

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service