C4105

Sigma-Aldrich

ColorBurst Electrophoresis Marker

pH 8, solution, Apparent Molecular Weight Range 8-220 kDa

MDL number:
Pricing and availability is not currently available.

form

solution

pH

8

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Brilliantly colored, exceptionally well-resolved, convenient, and stable, Colorburst protein molecular weight markers perform impressively in a variety of gel compositions and concentrations. ColorBurst Markers are composed of 8 polypeptides which have been chemically reduced and conjugated to brilliantly colored dyes. Conveniently, Colorburst Markers require no resuspension, reduction, or heating prior to use.

Application

Colorburst can be used to estimate sample molecular weights, to monitor the progress of an electrophoretic run, or to confirm that an electroblot is complete.
Colorburst can be used to estimate sample molecular weights, to monitor the progress of an electrophoretic run, or to confirm than an electroblot is complete.
Brilliantly colored, exceptionally well-resolved, convenient, and stable, ColorBurst protein molecular weight markers perform impressively in a variety of gel compositions and concentrations. ColorBurst Markers are composed of 8 polypeptides which have been chemically reduced and conjugated to brilliantly colored dyes. Conveniently, ColorBurst Markers require no resuspension, reduction, or heating prior to use.

Other Notes

Each vial of ColorBurst contains 500 μL of solution, enough for at least 50 mini gel applications.

Quantity

Each vial of ColorBurst contains 500 μL of solution, enough for at least 50 mini gel applications.

Legal Information

ColorBurst is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Codes

T

Risk Statement

61-40-48/22

Safety Statement

53-36/37-45

RIDADR

NONH for all modes of transport

WGK Germany

1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.