All Photos(1)

C5377

Sigma-Aldrich

Z-Trp-OH

Synonym(s):
Z-L-tryptophan, N-Cbz-L-tryptophan, Nα-Carbobenzyloxy-L-tryptophan
Empirical Formula (Hill Notation):
C19H18N2O4
CAS Number:
Molecular Weight:
338.36
Beilstein:
96743
EC Number:
MDL number:

mp

124-127 °C (lit.)

SMILES string

OC(=O)[C@H](Cc1c[nH]c2ccccc12)NC(=O)OCc3ccccc3

InChI

1S/C19H18N2O4/c22-18(23)17(10-14-11-20-16-9-5-4-8-15(14)16)21-19(24)25-12-13-6-2-1-3-7-13/h1-9,11,17,20H,10,12H2,(H,21,24)(H,22,23)/t17-/m0/s1

InChI key

AHYFYYVVAXRMKB-KRWDZBQOSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service