C5733

Sigma-Aldrich

Collagen from mouse sternum

Bornstein and Traub Type II, acid soluble, powder

CAS Number:
EC Number:
MDL number:
Pricing and availability is not currently available.

form

powder

impurities

Collagen Type I, free
<0.4% proteoglycan

solubility

acetic acid: 0.01-0.5 M

storage temp.

2-8°C

Gene Information

mouse ... Col1a1(12842)

General description

Collagen is classified into a number of structurally and genetically distinct types. We use the nomenclature proposed by Bornstein and Traub. Do not confuse Sigma type designations with recognized collagen classification types.

Application

Collagen-type 2 has been investigated for its importance in haemostasis. Two peptides, CB8 and CB11, are sites of platelet binding and activation on collagen-type 2. Collagen-type 2 has also been used as a model for arthritis in animals, where FR140423, a selective cyclooxygenase (COX)-2 inhibitor, has shown an anti-inflammatory effect without gastric lesions that may occur with treatment via indomethacin

Collagen Type II is sometimes used to induce an animal model of rheumatoid arthritis. However, under conditions leading to formation of reactive oxygen species, the subjects develop tolerance for auto-antibodies. This is an unusual situation of ROS having a favorable effect.

Biochem/physiol Actions

Acid soluble, lyophilized, salt-free, chromatographically purified powder from mouse sternum cartilage by modified methods

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

RIDADR

NONH for all modes of transport

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
T Ochi et al.
European journal of pharmacology, 365(2-3), 259-266 (1999-02-13)
The pharmacological profile of a novel and newly discovered non-steroidal anti-inflammatory and analgesic compound, 3-(difluoromethyl)-1-(4-methoxyphenyl)-5-[4-(methylsulfinyl)phenyl]pyraz ole (FR140423), was investigated. In recombinant human cyclooxygenase enzyme assays, the inhibition of prostaglandin E2 formation by FR140423 was 150 times more selective for cyclooxygenase-2...
Malin Hultqvist et al.
Journal of immunology (Baltimore, Md. : 1950), 179(3), 1431-1437 (2007-07-21)
The view on reactive oxygen species (ROS) in inflammation is currently shifting from being considered damaging toward having a more complex role in regulating inflammatory reactions. We recently demonstrated a role of ROS in regulation of animal models for the...
V K Il'in et al.
Vestnik Rossiiskoi akademii meditsinskikh nauk, (2)(2), 56-62 (2013-07-04)
Dysbiotic shifts in intestinal and pharyngeal microflora were studied in 22 normal volunteers in 9-, 14-, 105- and 520-d chamber experiments simulating some of the spaceflight factors. Two preparations were administered to prevent pharyngeal and intestinal dysbiosis: oral dry probiotic...
Michelle K Nguyen et al.
Current opinion in ophthalmology, 24(4), 291-295 (2013-06-06)
To describe the use of corneal collagen cross-linking (CXL) and its efficacy in the stabilization of keratorefractive procedures, including PRK, laser in-situ keratomileusis (LASIK), thermal keratoplasty, and orthokeratology. Since its introduction, CXL has quickly gained interest in the treatment of...
V V Vereshchaka
Fiziolohichnyi zhurnal (Kiev, Ukraine : 1994), 59(1), 97-109 (2013-05-30)
It was examined 120 persons of both sexes who had some problems with face skin. It was investigated the degree of face age changes in persons with the symptoms of skin slacses and practically healthy persons of different ages. Some...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.