C5770

Sigma-Aldrich

Z-Gly-Gly-Arg 4-methoxy-β-naphthylamide hydrochloride

Empirical Formula (Hill Notation):
C29H35N7O6 · HCl
CAS Number:
Molecular Weight:
614.09
MDL number:
PubChem Substance ID:
Pricing and availability is not currently available.

form

solid

storage temp.

−20°C

SMILES string

Cl.COc1cc(NC(=O)C(CCCNC(N)=N)NC(=O)CNC(=O)CNC(=O)OCc2ccccc2)cc3ccccc13

InChI

1S/C29H35N7O6.ClH/c1-41-24-15-21(14-20-10-5-6-11-22(20)24)35-27(39)23(12-7-13-32-28(30)31)36-26(38)17-33-25(37)16-34-29(40)42-18-19-8-3-2-4-9-19;/h2-6,8-11,14-15,23H,7,12-13,16-18H2,1H3,(H,33,37)(H,34,40)(H,35,39)(H,36,38)(H4,30,31,32);1H

InChI key

ZHYMKOBIONUEST-UHFFFAOYSA-N

Substrates

Fluorogenic substrate for urokinase.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

Xn

Risk Statement

40

Safety Statement

22-36

RIDADR

NONH for all modes of transport

WGK Germany

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin
Studies on tissue culture plasminogen activator. II. The detection and assay of urokinase and plasminogen activator from LLC-PK cultures (porcine) by the synthetic substrate Nalpha-benzyloxycarbonyl-glycyl-glycyl-arginyl-4-methoxy-2-napthylamide.
R M Huseby et al.
Thrombosis research, 10(5), 679-687 (1977-05-01)
Sensitive fluorescence assays for urokinase using synthetic peptide 4-methoxy-beta-naphthylamide substrates.
W L Bigbee et al.
Analytical biochemistry, 88(1), 114-122 (1978-07-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.