MilliporeSigma
All Photos(1)

C6112

Sigma-Aldrich

CD30 Ligand human

>95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym(s):
CD153, CD30L
MDL number:

recombinant

expressed in NSO cells

assay

>95% (SDS-PAGE)

form

lyophilized powder

mol wt

30-40 kDa by SDS-PAGE (reducing)
calculated mol wt 22 kDa

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... TNFSF8(944)

Biochem/physiol Actions

CD30 ligand, a type II membrane protein, is a member of the TNF family. The specific receptor for CD30L is CD30. CD30-CD30L interaction mediates cell proliferation, cell activation, cell differentiation, and cell death.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Analysis Note

The biological activity is measured by its ability to stimulate IL-6 secretion by HDLM cells, a Hodgkin′s lymphoma cell line.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service