C8301

Sigma-Aldrich

Sigma Blend Collagenase Type N from Clostridium histolyticum

<0.1 FALGPA units/mg protein (Neutral Protease: 80-120 units per mg solid)

EC Number:
Pricing and availability is not currently available.

specific activity

<0.1 FALGPA units/mg protein (Neutral Protease: 80-120 units per mg solid)

storage temp.

−20°C

Legal Information

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Target Organs

Respiratory system

Personal Protective Equipment

dust mask type N95 (US),Eyeshields,Gloves

Hazard Codes

Xn

Risk Statement

36/37/38-42

Safety Statement

22-24-26-36/37

RIDADR

NONH for all modes of transport

WGK Germany

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.