Documents

CS0070

Sigma-Aldrich

p53 ELISA human

sufficient for 96 multiwell tests

Synonym(s):
p53 ELISA
Pricing and availability is not currently available.

usage

sufficient for 96 multiwell tests

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... TP53(7157)

General description

A solid phase sandwich enzyme-linked immunosorbent assay (ELISA).

Application

p53 ELISA is designed to detect and quantify the levels of human p53 protein, independent of its phosphory­lation state. It is used to provide the total p53 levels in the samples, which are simultaneously tested for p53 phosphorylated at serine 15.
Designed for the in vitro quantitative determination of human p53 protein in cell lysates. The assay does not react with p53 in mouse and rat.

Biochem/physiol Actions

p53 gene is highly conserved and expressed in normal tissues. It is the most commonly mutated gene in human cancer and more then 500 gene mutations have been described in various types of malignancies, hematologic as well as solid tumors. Intact p53 function is essential for the maintenance of the non-tumorogenic phenotype of cells. Thus, p53 plays a vital role in suppressing the development of cancer.

Features and Benefits

p53 ELISAs offer a fast (total assay time 4 hours), sensitive (<50 pg/mL for total p53 and <0.9 units/mL for phosphospecific p53, which is 8x more sensitive than immunoblotting), and an easy alternative to immunoblotting or other bioassays.

Analysis Note

The sensitivity is <50 pg/mL for total p53, which is 8x more sensitive than immunoblotting.

Safety Statement

24/25

RIDADR

UN 3316 9/PG 3

Certificate of Analysis
Certificate of Origin
G J Milczarek et al.
Life sciences, 60(1), 1-11 (1997-01-01)
The p53 protein plays a vital role in suppressing the development of cancer. Posttranslational modification through phosphorylation has been postulated to be an important regulatory mechanism of p53 function. Data describing the role of phosphorylation in terms of its effects...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.