Documents

CS0100

Sigma-Aldrich

JNK 1/2 ELISA

sufficient for 96 tests

Synonym(s):
c-Jun N-terminal Kinase (JNK)1&2 ELISA
Pricing and availability is not currently available.

usage

sufficient for 96 tests

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... MAPK8(5599), MAPK9(5601)

Looking for similar products? Visit Product Comparison Guide

General description

JNK1&2 ELISAs are designated for quantitative determination of human and mouse phosphorylated or non-phosphorylated JNK1&2 in cell lysates. The antibodies in the JNK1&2 ELISAs require that the protein in cells be denatured in order to achieve recognition. The assay is a solid-phase sandwich enzyme-linked immunosorbent assay (ELISA). A monoclonal capture antibody specific for the JNK1&2 has been coated onto the multiwell strips provided with the kit. Standard dilutions and samples are incubated for 2 hours at room temperature. JNK1&2 antigen binds to the capture antibody. After a wash, a detection antibody specific for the non-phosphorylated or phosphorylated JNK1&2 protein is incubated for 1 hr at room temperature, which results in binding of the antibody to the immobilized JNK1&2 protein. An anti-rabbit IgG-HRP binds to the immobilized protein, completing the four-member sandwich. The reaction is visualized by tetramethylbenzidine (TMB) substrate, followed by the stop solution. The intensity of the yellow color, measured in a multiwell plate reader at 450 nm, is directly proportional to the concentration of JNK1&2 in the original sample. The unknown concentrations are calculated from the standard curve run with each assay.

Biochem/physiol Actions

JNK plays a role in apoptosis and cell survival. JNK1 and JNK2 are required for polarized differentiation of T-helper cells into Th1 cells. Activation of JNK1&2 causes phosphorylation of a number of transcription factors, such as c-Jun and ATF2, and thus regulates AP-1 transcriptional activity.

Features and Benefits

JNK1&2 ELISAs are fast (total time 4 hours), sensitive (detects <0.15 ng/mL or <0.8 units/mL), and specific for either non-phosphorylated or phosphorylated JNK1&2. Precoated plates are provided. All incubations are at room temperature and no proprietary equipment is required.

Analysis Note

JNK1&2 ELISA is specific for native and recombinant human and mouse JNK1&2 regardless of phosphorylation state. The sensitivity is <0.15 ng/mL and equals the sensitivity of immunobloting. Linear regression yielded a correlation coefficient of 0.99.

RIDADR

UN 3316 9/PG 2

Certificate of Analysis
Certificate of Origin
Y Fleming et al.
The Biochemical journal, 352 Pt 1, 145-154 (2000-11-04)
Stress-activated protein kinase 1 (SAPK1), also called c-Jun N-terminal kinase (JNK), becomes activated in vivo in response to pro-inflammatory cytokines or cellular stresses. Its full activation requires the phosphorylation of a threonine and a tyrosine residue in a Thr-Pro-Tyr motif...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.