Documents

CS0180

Sigma-Aldrich

Substance P EIA

sufficient for 96 multiwell tests

MDL number:
Pricing and availability is not currently available.

usage

sufficient for 96 multiwell tests

shipped in

dry ice

storage temp.

−20°C

Biochem/physiol Actions

Substance P is an 11 amino acid (H-Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2) peptide that is synthesized in ribosomes as a larger protein and then enzymatically converted into the active peptide. The peptide belongs to the tachykinin family and is widely distributed in the peripheral and central nervous systems of vertebrates, where it acts as a neurotransmitter. Substance P is involved in pain, anxiety, depression and inflammation.
Substance P, widely distributed in the peripheral and central nervous systems, acts as a neurotransmitter involved in pain, anxiety, depression, and inflammation.

Features and Benefits

Substance P EIA is fast (total incubation time 3 hours), specific (measures levels with no cross-reactivity with neurokinins and somatostatins), and sensitive (sensitivity less than 8.04 pg/mL). All components are stable at 2-8 ºC. Sample recovery for various matrices, ranges from 81% (cell culture supernatants diluted 1:2) to 100% (porcine heparin plasma). High assay precision, intra-assay CV 5.2-6.7% and inter-assay CV 4.2-7.3%.

Hazard Codes

C

Risk Statement

22-34

Safety Statement

26-36/37/39-45

RIDADR

UN 3316 9/PG 3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.