Documents

CS0310

Sigma-Aldrich

Caspase 10 Fluorescein (FLICA) Assay

sufficient for 100 tests

Synonym(s):
Caspase Fluorescein (FLICA) Assay
Pricing and availability is not currently available.

usage

sufficient for 100 tests

storage temp.

2-8°C

Application

Fluorometric detection of active cellular caspase 10 using a cell-permeable, non-toxic FAM-AEVD-FMK fluorochrome inhibitor of caspase 10 that covalently binds to the enzyme and is visualized by fluorometry, fluorescence microscopy, or flow cytometry.
FLICA Caspase Assay detects active caspases inside the cell using novel Fluorochrome Inhibitor of Caspases (FLICA)-cell permeable and non-cytotoxic caspase inhibitors. Once inside the cell, the FLICA probe covalently binds to a reactive cysteine residue on the large subunit of the active caspase heterodimer, thereby inhibiting further enzymatic activity. Bound FLICA is retained within the cell, while any unbound reagent is washed away. The bound FLICA emits a green fluorescent signal, which is a direct measurement of the number of active caspase molecules that are present in the cell at the time the reagent was added.

Features and Benefits

The FLICA Assays offer:
  • Potent, cell-permeable and non-toxic fluorochrome inhibitor
  • Preferential detection of selected caspases based on inhibitor specificity
  • A direct measure of apoptosis expressed as the number of active caspase enzymes present in the cell

Caution

Once reconstituted, the unused 150X FLICA stock should be stored in aliquots at −20 °C protected from light.

Kit Components Only

Product No.
Description

  • FAM-AEVD-FMK FLICA Reagent 4 vial(s)

  • Wash Buffer, Concentrate 10X 60 mL

  • Fixative 6 mL

  • Propidium iodide 1 mL

  • Hoechst Stain 33342 1 mL

Hazard Codes

T

Risk Statement

5-23/24/25-32-40-42/43-52/53

Safety Statement

23-36/37-45

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.