Documents

CS0360

Sigma-Aldrich

Cathepsin Detection Assay

Cathepsin K Detection Assay, sufficient for 100 assays

Pricing and availability is not currently available.

usage

sufficient for 100 assays

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cathepsins are apoptosis markers associated with Alzheimer′s disease, numerous types of cancer, autoimmune arthritis, and the breakdown of bone structure seen with osteoporosis.

Principle

Cathepsin K detection assay uses (z-Leucine-Arginine)2(z-LR)2 derivative of the cresyl violet fluorophore, which easily penetrates the cell membrane and the membranes of the internal cellular organelles enabling detection of cathepsin K activity within whole living cells. Hoechst stain is used to label the cell nuclei after labeling with the MR- Cathepsin K substrate. It is visualized under a microscope using a UV-filter with excitation at 365 nm and emission at 480 nm. Acridine orange (AO) stains aggregates in acidic pH and fluoreses orange rather than green, thus clearly differentiating the lysosomes from the other organelles.
Cathepsin B, K, and L protease activities can be detected within whole living cells using substrate-based fluorescent assays, which utilize cathepsin target sequence peptides, derivatives of the cresyl violet fluorophore, conjugated to Magic Red™ (MR) fluorogenic substrate (cresyl violet). Following enzymatic cleavage inside the living cells at one or both arginine (R) amide linkage sites, the mono and non-substituted cresyl violet fluorophores generate red fluorescence when excited at 550-590 nm.

Legal Information

Magic Red™ trademark is owned by MP Biomedicals (Formerly ESP).

Kit Components Only

Product No.
Description

  • MR-(LR)2 substrate 1 mL/vial

  • Hoechst 33342 stain 1 mL

  • Acridine orange stain (AO) .5 mL

Certificate of Analysis
Certificate of Origin
G Motyckova et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(10), 5798-5803 (2001-05-02)
Various genetic conditions produce dysfunctional osteoclasts resulting in osteopetrosis or osteosclerosis. These include human pycnodysostosis, an autosomal recessive syndrome caused by cathepsin K mutation, cathepsin K-deficient mice, and mitf mutant rodent strains. Cathepsin K is a highly expressed cysteine protease...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.