Documents

CS0440

Sigma-Aldrich

phospho-Tau (pSer199) ELISA mouse

sufficient for 96 determinations

Synonym(s):
Tau ELISA
Pricing and availability is not currently available.

usage

sufficient for 96 determinations

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... MAPT(4137)

General description

A solid phase sandwich enzyme-linked-immunosorbent assay (ELISA).

Application

The assay is a solid phase sandwich enzyme linked-immunosorbent assay (ELISA) for the in vitro quantitative determination of mouse Tau in mouse brain homogenates, cell extracts, buffered solutions, or cell culture media. The assay will recognize both natural and recombinant mTau.
The assay uses a monoclonal capture antibody specific for mouse tau that has been coated onto multiwell strips. Standard dilutions and samples are incubated 2 hours at RT. Tau antigen binds to the capture antibody. After a wash, a detection antibody specific for the non-phosphorylated or phosphorylated protein is incubated 1 hr at RT, which results in binding to the immobilized Tau protein. An anti-rabbit IgG-HRP binds to the immobilized protein completing the four-member sandwich. The reaction is visualized by tetramethylbenzidine (TMB) substrate, followed by the stop solution. The intensity of the yellow color measured in a multiwell plate reader at 450 nm is directly proportional to the concentration of Tau in original sample. The unknown concentrations are calculated from the standard curve run with each assay

Features and Benefits

Tau ELISA is a simple, sensitive, and specific alternative to immunoblotting and RIA.

Analysis Note

The specificity was confirmed by peptide competition. The data shows that only the peptide corresponding to the region surrounding serine 199 containing the phosphoserine blocks the ELISA signal.

Kit Components Only

Product No.
Description

  • Phospho-Mouse Tau (pSer199) Standard 2 vial(s)

Hazard Codes

T

Risk Statement

23-34

Safety Statement

26-36/37/39-45

RIDADR

UN 3316 9/PG 3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.