Documents

CS0470

Sigma-Aldrich

STAT1 ELISA human

sufficient for 96 determinations

Synonym(s):
STAT1 ELISA
Pricing and availability is not currently available.

usage

sufficient for 96 determinations

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... STAT1(6772)

General description

A solid phase sandwich enzyme-linked immunosorbent assay (ELISA).

Application

The assay detects and quantifies the level of STAT1 protein from lysates of human cells or tissues independent of its phosphorylation state. It recognizes both α and β isoforms of STAT1. It does not detect STAT1 from mouse or rat cells. The assay may be used to normalize the STAT1 content of the samples when examining quantities of phosphorylated sites on STAT1 using another Sigma kit Phospho-STAT1 pTyr701 ELISA (CS0480).
The assay is designed to detect and quantify the level of STAT1.

Biochem/physiol Actions

STAT1, a member of the STAT family of proteins, controls the transcription of specific genes in response to cytokine stimulation. STAT1 mediates apoptotic signals from interferon γ stimulation and may modulate anti-apoptotic signaling arising from NF-κB.

Features and Benefits

STAT1 ELISA is a fast (total time 4 hours), sensitive, and specific alternative to immuniblotting and RIA.

Analysis Note

The sensitivity is <0.27 ng/mL of STAT1, which is 4x more sensitive than immunoblotting. Linear regression yielded a correlation coefficient of 0.99.

Kit Components Only

Product No.
Description

  • STAT1 Standard 2 vial(s)

  • Standard Diluent Buffer 25 mL

  • Monoclonal-Anti-STAT1-Coated 96 well plate 1 ea

  • Anti-STAT1 11 mL

  • Anti-Rabbit IgG-HRP 1 vial

  • Sample Treatment Buffer 10 mL

  • HRP Diluent 25 mL

  • Wash Buffer concentrate, 25X 100 mL

  • Stabilized Chromogen, Tetramethylbenzidine (TMB) 25 mL

  • Stop Solution 25 mL

  • Plate Covers, Adhesive strips 3 ea

See All (11)

Safety Statement

23-24/25

RIDADR

UN 3316 9/PG 3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.