Documents

CS0520

Sigma-Aldrich

Bcl-2 ELISA human

sufficient for 96 determinations

Synonym(s):
Bcl-2 ELISA
Pricing and availability is not currently available.

usage

sufficient for 96 determinations

shipped in

wet ice

storage temp.

2-8°C

General description

The assay is a solid-phase sandwich enzyme-linked immunosorbent assay (ELISA). Standard dilutions and samples are added to the wells precoated with Anti-Bcl-2 antibody (regardless of phosphorylation state)and incubated 2 hours at RT. Bcl-2 binds to the capture antibody. A detection antibody specific for the non-phosphorylated or phosphorylated protein isadded and incubated 1 hr at RT, which results in binding to the immobilized IR protein. An anti-rabbit IgG-HRP binds to the immobilized protein completing the four-member sandwich. The reaction is visualized by tetramethylbenzidine (TMB) substrate, followed by the stop solution. The intensity of the yellow color, measured in a multiwell plate reader at 450 nm, is directly proportional to the concentration of Bcl-2 in standards and samples. The unknown concentrations are calculated from the standard curve run with each assay.

Application

Bcl-2 ELISA is designed to detect and quantify the level of human Bcl-2 protein in cell lysates. Human Bcl-2, the mammalian homolog of the Caenorhabditis elegans protein CED-9, is a cellular oncoprotein with MW 26 kDa, which localizes to the mitochondrial outer membrane, the endoplasmic reticulum, and the nuclear membrane. Bcl-2 has two functions in the cell: to promote cell survival, and to encourage cell cycle exit and quiescence.
The assay is specific for human Bcl-2 protein regardless of phosphorylation status. p53, Akt, and Src show no cross-reactivity.There is no reactivity with mouse or rat Bcl-2.

Features and Benefits

Bcl-2 ELISA is a fast (total time 4 hours), sensitive, and specific alternative to immunoblotting and RIA. The ease of run is facilitated by precoated plates and ready-to-use reagents.

Analysis Note

Bcl-2 ELISA is a sensitive and specific alternative to immunoblotting and RIA.

Kit Components Only

Product No.
Description

  • Bcl-2 Standard, Lyophilized 2 vial(s)

  • Standard Diluent Buffer 25 mL

  • Monoclonal-Anti-Bcl-2 Coated 96 wells plate 1 ea

  • Anti-Bcl-2 11 mL

  • Anti-Rabbit IgG-HRP, Concentrate (100X 1 vial

  • HRP Diluent 25 mL

  • Wash Buffer concentrate, 25X 100 mL

  • Stabilized Chromogen, Tetramethylbenzidine (TMB), 25 mL

  • Stop Solution 25 mL

  • Plate Covers, Adhesive strips, 3 ea

See All (10)

RIDADR

UN 3316 9/PG 3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.