Documents

CS0550

Sigma-Aldrich

phospho-ATF2 (pThr69/pThr71) ELISA human

sufficient for 96 determinations

Synonym(s):
ATF2 ELISA, Activating Transcription Factor 2
Pricing and availability is not currently available.

usage

sufficient for 96 determinations

shipped in

wet ice

storage temp.

2-8°C

General description

ATF2 ELISA is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (ELISA) for quantitative determination of ATF2 protein in cell lysates. ATF2 standard dilutions, controls, and unknown samples are added to wells precoated with monoclonal antibody specific for ATF2 (regardless of phosphorylation state) and incubated 2 hours at RT. ATF2 antigen binds to the capture) antibody. Detection Anti- ATF2 antibody is added to the wells and binds to the immobilized ATF2. Anti-IgG- HRP binds to the detection antibody to complete the four-member sandwich. A substrate solution is added, which is acted upon by the bound enzyme to produce color. The intensity of this colored product is directly proportional to the concentration of ATF2 in the original specimen. The optical density measured at 450 nm in the multiwell plate reader is used to calculate the concentration of ATF2

Application

ATF2 ELISA detects and quantifies the level of ATF2 protein. Performance characterization of this ELISA kit was done primarily on human cell lines, cross-reactivity of this kit with mouse cells was observed. ATF2 (activating transcription factor 2) is a member of the ATF/CREB (cAMP response element-binding protein) family of basic region-leucine zipper proteins. Activation of ATF2 is observed in cellular responses to various types of stress and apoptotic signals
Phospho-ATF2 pThr69/pThr71 ELISA is designed to detect and quantify the level of ATF2 protein double phosphorylated on threonines 69 and 71. Although performance characterization of this ELISA was done primarily on human cell lines, this kit cross-reacts with mouse and rat cells. To normalize the ATF2 content of the samples when examining quantities of phosphorylated sites on ATF2 use Sigma ATF2 ELISA (Product No. CS0540).

Analysis Note

The sensitivity is <1 units/mL and 8x more sensitive than immunoblotting.

Kit Components Only

Product No.
Description

  • Phospho-ATF2 pThr69/pThr71 Standard, Lyophilized 2 vial(s)

  • Standard Diluent Buffer 25 mL

  • Monoclonal-Anti-ATF2-Coated 96 well plate 1 ea

  • Anti-phospho-ATF2 pThr69/pThr71 11 mL

  • Anti-Rabbit IgG-HRP, Concentrate (100X 1 vial

  • HRP Diluent 25 mL

  • Wash Buffer concentrate, 25X 100 mL

  • Stabilized Chromogen, Tetramethylbenzidine (TMB) 25 mL

  • Stop Solution 25 mL

  • Plate Covers, Adhesive strips 3 ea

See All (10)

RIDADR

UN 3316 9/PG 3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.