Documents

CS0630

Sigma-Aldrich

phospho-Heat Shock Protein 27 (pSer82) ELISA human

sufficient for 96 determinations

Synonym(s):
HSP27, Heat Shock Protein 27 ELISA
MDL number:
Pricing and availability is not currently available.

usage

sufficient for 96 determinations

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... HSPB1(3315)

General description

A solid phase sandwich enzyme-linked immunosorbent assay (ELISA). Standard dilutions and samples are added to the wells precoated with Anti-HSP27 capture antibody, specific for HSP27 regardless of phosphorylation state. After 2 hr at RT, a detection antibody specific for the non-phosphorylated or phosphorylated protein is added and incubated 1 hr at RT, which results in binding to the immobilized HSP27 protein. An anti-rabbit IgG-HRP binds to the immobilized protein completing the four-member sandwich. The reaction is visualized by TMB substrate, followed by the stop solution. The intensity of the yellow color, measured in a multiwell plate reader at 450 nm, is directly proportional to the concentration of HSP27 in original sample. The unknown concentrations are calculated from the standard curve run with each assay.

Application

Phospho-Heat Shock Protein 27 pSer82 ELISA is designed to detect and quantify the level of HSP27 protein, phosphorylated on serine 82. This assay is intended for the detection of Phospho-HSP27 pSer82 in lysates of human cells. It does not react with the mouse or rat HSP27 homolog, HSP25. For normalizing the HSP27 content of the samples, Heat Shock Protein 27 ELISA, which permits quantification of HSP27 independent of phosphorylation status, is available from Sigma (Product No. CS0640).

Biochem/physiol Actions

HSP27, a member of the ubiquitously expressed anti-apoptotic heat shock protein group, plays a role in maintaining cellular redox by increasing glutathione levels, stabilizing actin filaments, and blocking actin polymerization. It prevents the aggregation of improperly folded proteins. Recent evidence suggests that HSP27 protects cells from apoptosis by participation in several signaling pathways.

Analysis Note

The sensitivity is <1.0 units/mL, which is 4 times more than immunoblotting. The specificity is confirmed by peptide competition. Only the phosphopeptide blocks the ELISA signal.

Kit Components Only

Product No.
Description

  • Phospho-HSP27 pSer82 Standard, Lyophilized 2 vial(s)

  • Standard Diluent Buffer 25 mL

  • Monoclonal-Anti-HSP27-Coated 96 well plate 2 vial(s)

  • Anti-Phospho-HSP27 pSer82 6 mL

  • Anti-Rabbit IgG-HRP, Concentrate (100X) 1 vial

  • HRP Diluent 25 mL

  • Wash Buffer concentrate, 25X 100 mL

  • Stabilized Chromogen (TMB) 25 mL

  • Stop Solution 25 mL

  • Plate covers, Adhesive Strips 3 ea

See All (10)

Safety Statement

23-24/25

RIDADR

UN 3316 9/PG 3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.