Documents

CS0700

Sigma-Aldrich

MEK1 ELISA

sufficient for 96 determinations

Synonym(s):
MEK1 ELISA, ERK kinase 1, MAPK kinase 1
Pricing and availability is not currently available.

usage

sufficient for 96 determinations

shipped in

wet ice

storage temp.

2-8°C

Gene Information

human ... MAP2K1(5604)

General description

The assay is a solid-phase sandwich enzyme-linked immunosorbent assay (ELISA). A capture antibody specific for MEK1 regardless of phosphorylation status, has been coated onto the multiwell strips. Standard dilutions and samples are incubated 2 hours at RT and MEK1 antigen binds to the capture antibody. After wash, a detection antibody specific for the non-phosphorylated or phosphorylated protein is incubated 1 hr at RT, and binds to the immobilized MEL1 protein. An anti-rabbit IgG-HRP completes the four-member sandwich. The reaction is visualized by TMB substrate, followed by the stop solution. The intensity of the yellow color, measured in a multiwell plate reader at 450 nm, is directly proportional to the concentration of MEK1 in original sample. The unknown concentrations are calculated from the standard curve run with each assay
Use the multiwell plate and components incuded in the assay, following instruction provided in Technical Bulletin. Components are lot-specific and not interchangeable.

Application

MEK1 ELISA is designed to detect and quantify the level of MEK1 protein independent of its phosphorylation state from lysates of human, mouse, and rat cells.

Biochem/physiol Actions

MEK1, also known as ERK kinase 1, MAPK kinase 1, and MKK1, is a member of the MEK family of dual specificity protein kinases.

Features and Benefits

MEK1 ELISAs are a fast (total time 4 hours), sensitive, and specific alternative to immuniblotting and RIA. The assay is facilitated by precoated plates and ready-to-use reagents.

Analysis Note

The sensitivity is <0.1 ng/mL of MEK1, which is 2 times more sensitive than immunoblotting. Linear regression yielded a correlation coefficient of 0.99.

Kit Components Only

Product No.
Description

  • MEK1 Standard, Lyophilized 2 vial(s)

  • Standard Diluent Buffer 25 mL

  • Sample Treatment Buffer 10 mL

  • Monoclonal-Anti-MEK1 Coated 96 well plate 1 ea

  • Anti-MEK1 5.5 mL

  • Anti-Rabbit IgG-HRP, Concentrate (100X 1 vial

  • HRP Diluent 25 mL

  • Wash Buffer concentrate, 25X, 100 mL

  • Stabilized Chromogen TMB 25 mL

  • Stop Solution 25 mL

  • Plate Covers, Adhesive strips 3 ea

See All (11)

RIDADR

UN 3316 9/PG 3

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.