Documents

CYTO1A

Sigma-Aldrich

Cytochrome P450 1A Fluorescent Detection Kit

Pricing and availability is not currently available.

storage temp.

−20°C

Biochem/physiol Actions

Cytochrome P4501A1 and P4501A2 activity is induced in mammalian liver by compounds such as β-naphthoflavone, benzoapyrene, 3-methylcholanthrene and 2,3,7,8-tetrachlorodibenzo-p-dioxin.

Packaging

Ethoxyresorufin and methoxyresorufin are supplied as substrates.

Hazard Codes

Xi

Risk Statement

36/37/38

Safety Statement

26

Certificate of Analysis
Certificate of Origin
X Rey-Grobellet et al.
Xenobiotica; the fate of foreign compounds in biological systems, 26(7), 765-778 (1996-07-01)
1. Thiabendazole is a widely used food preservative and anthelmintic drug for breeding animal species. In order to characterize precisely the cytochrome P450 isozyme(s) involved in its major route of metabolism, a rapid and sensitive spectrofluorimetric method was developed for...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.