Documents

CYTO2B

Sigma-Aldrich

Cytochrome P450 2B Fluorescent Detection Kit

Pricing and availability is not currently available.

storage temp.

−20°C

Biochem/physiol Actions

Cytochrome P450 2B activity is induced in mammalian liver by compounds such as chlorinated hydrocarbons, aromatics and barbiturates.

Packaging

Pentoxyresorufin is supplied as substrate.

Hazard Codes

Xi

Risk Statement

36

Safety Statement

26

Certificate of Analysis
Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service

Social Media

LinkedIn icon
Twitter icon
Facebook Icon
Instagram Icon

MilliporeSigma

Research. Development. Production.

We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.

© 2021 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved.

Reproduction of any materials from the site is strictly forbidden without permission.