All Photos(1)

D1290

Supelco

1-Deoxyforskolin from Coleus forskohlii

≥98%

Synonym(s):
7β-Acetoxy-6β,9α-dihydroxy-8,13-epoxy-labd-14-en-11-one
Empirical Formula (Hill Notation):
C22H34O6
CAS Number:
Molecular Weight:
394.50
MDL number:
PubChem Substance ID:

assay

≥98%

form

neat

application(s)

HPLC: suitable
gas chromatography (GC): suitable

Featured Industry

Food and Beverages

format

neat

storage temp.

2-8°C

SMILES string

O=C1C[C@](C)(C=C)O[C@]2(C)[C@@H](OC(C)=O)[C@@H](O)[C@@]3([H])C(C)(C)CCC[C@]3(C)[C@]21O

InChI

1S/C22H34O6/c1-8-19(5)12-14(24)22(26)20(6)11-9-10-18(3,4)16(20)15(25)17(27-13(2)23)21(22,7)28-19/h8,15-17,25-26H,1,9-12H2,2-7H3/t15-,16-,17-,19-,20-,21+,22-/m0/s1

InChI key

PXYAFNGUEZPJBI-OAUGPMCWSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Certificate of Analysis

Certificate of Origin

Khandelwal, Y., et al.
Tetrahedron Letters, 45, 763-763 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service