All Photos(1)

D5759

Sigma-Aldrich

2,4-Dimethylthiazole

Empirical Formula (Hill Notation):
C5H7NS
CAS Number:
Molecular Weight:
113.18
EC Number:
MDL number:
PubChem Substance ID:

refractive index

n20/D 1.509 (lit.)

bp

144-146 °C/719 mmHg (lit.)

density

1.056 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

Cc1csc(C)n1

InChI

1S/C5H7NS/c1-4-3-7-5(2)6-4/h3H,1-2H3

InChI key

OBSLLHNATPQFMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service