MilliporeSigma
All Photos(1)

D6939

Sigma-Aldrich

Delcorine

≥97% (TLC), solid

Empirical Formula (Hill Notation):
C26H41NO7
Molecular Weight:
479.61
MDL number:

assay

≥97% (TLC)

form

solid

storage temp.

2-8°C

SMILES string

CCN1C[C@@]2(CC[C@H](OC)[C@@]34[C@@H]5C[C@@H]6[C@H](C[C@@]7(OCO[C@]7([C@@H](O)[C@H]23)[C@@H]14)[C@H]5[C@H]6OC)OC)COC

Biochem/physiol Actions

Antiarrhythmic and hypotensive; ganglion blocker; exhibits weak affinity at brain α-bungarotoxin-sensitive nicotinic acetylcholine receptors.

Caution

Light sensitive.

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More documents

Quotes and Ordering

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service