All Photos(1)

D8390

Sigma-Aldrich

1,4-Dideoxy-1,4-imino-D-mannitol hydrochloride

≥90% (HPLC)

Empirical Formula (Hill Notation):
C6H13NO4 · HCl
CAS Number:
Molecular Weight:
199.63
MDL number:
PubChem Substance ID:

assay

≥90% (HPLC)

form

solid

storage temp.

2-8°C

SMILES string

Cl.OC[C@@H](O)[C@H]1NC[C@@H](O)[C@H]1O

InChI

1S/C6H13NO4.ClH/c8-2-4(10)5-6(11)3(9)1-7-5;/h3-11H,1-2H2;1H/t3-,4-,5-,6-;/m1./s1

InChI key

IFRNNJQXHHLGKS-MVNLRXSJSA-N

Certificate of Analysis

Certificate of Origin

Inhibitors of the biosynthesis and processing of N-linked oligosaccharide chains.
A D Elbein
Annual review of biochemistry, 56, 497-534 (1987-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service