All Photos(1)

D8505

Sigma-Aldrich

2,6-Dithiopurine

Synonym(s):
2,6-Purinedithiol, Dithioxanthine, 2,6-Dimercaptopurine
Empirical Formula (Hill Notation):
C5H4N4S2
CAS Number:
Molecular Weight:
184.24
EC Number:
MDL number:

mp

>350 °C (lit.)

storage temp.

−20°C

SMILES string

Sc1nc(S)c2[nH]cnc2n1

Looking for similar products? Visit Product Comparison Guide

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service