All Photos(1)

E8377

Sigma-Aldrich

Elaidic acid ethyl ester

liquid

Empirical Formula (Hill Notation):
C20H38O2
CAS Number:
Molecular Weight:
310.51
EC Number:
MDL number:
PubChem Substance ID:

form

liquid

functional group

ester

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C\CCCCCCCC(=O)OCC

InChI

1S/C20H38O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h11-12H,3-10,13-19H2,1-2H3/b12-11+

InChI key

LVGKNOAMLMIIKO-VAWYXSNFSA-N

Packaging

Sealed ampule.

Certificate of Analysis

Certificate of Origin

Clarissa Hamaio Okino-Delgado et al.
PloS one, 12(10), e0186246-e0186246 (2017-10-27)
Cooking oil waste leads to well-known environmental impacts and its bioremediation by lipase-based enzymatic activity can minimize the high cytotoxic potential. In addition, they are among the biocatalysts most commercialized worldwide due to the versatility of reactions and substrates. However...
Soo J Park et al.
Molecules (Basel, Switzerland), 25(18) (2020-09-17)
The cuticular layer of the insect exoskeleton contains diverse compounds that serve important biological functions, including the maintenance of homeostasis by protecting against water loss, protection from injury, pathogens and insecticides, and communication. Bactrocera tryoni (Froggatt) is the most destructive...
Shujauddin Changi et al.
ChemSusChem, 5(9), 1743-1757 (2012-08-29)
We examined the behavior of phenylalanine in high-temperature water (HTW) at 220, 250, 280, and 350 °C. Under these conditions, the major product is phenylethylamine. The minor products include styrene and phenylethanol (1-phenylethanol and 2-phenylethanol), which appear at higher temperatures and...
Claudia Dussaubat et al.
Journal of chemical ecology, 36(5), 522-525 (2010-04-20)
Pheromones in social insects play a key role in the regulation of group homoeostasis. It is well-established that parasites can modify hormone signaling of their host, but less is known about the effect of parasites on pheromone signaling in insect...
Y Ingrid Goh et al.
Alcohol (Fayetteville, N.Y.), 44(7-8), 629-634 (2010-06-30)
Meconium fatty acid ethyl esters (FAEEs) are sensitive and specific biomarkers for prenatal alcohol exposure (PAE) in pregnancy. We recently reported a 2.5% rate of FAEE positive meconium in a general population sample of infants born in the region of...

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service