All Photos(1)

F1523

Sigma-Aldrich

Iron(III) phosphate

suitable for insect cell culture, BioReagent

Synonym(s):
Ferric phosphate
Linear Formula:
FePO4
CAS Number:
Molecular Weight:
150.82
MDL number:
PubChem Substance ID:

product line

BioReagent

technique(s)

cell culture | insect: suitable

SMILES string

[Fe+3].[O-]P([O-])([O-])=O

General description

Iron(II) sulfate heptahydrate is used in cell culture applications generally bound to transferrin, citrate or other iron transport molecules.

Other Notes

Compound is a mixture of ferric phosphate and sodium citrate.

Certificate of Analysis

Certificate of Origin

Sophie Rodrigues et al.
Probiotics and antimicrobial proteins, 7(1), 45-51 (2014-10-22)
Vibrio tapetis CECT4600 is a pathogenic Gram-negative bacterium causing the brown ring disease in the Manila clam Ruditapes philippinarum. This vibriosis is induced by bacterial attachment on the periostracal lamina, yielding a decalcification of the bivalve shell. As in many

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service